Gene Transcripts

Gene Symbol
IGSF23
Entrez Gene ID
147710
Gene Name
immunoglobulin superfamily member 23
Gene Aliases
Chromosome Location
Chr. 19: 44612577 - 44636781 on Build GRCh38
UniGene ID
Hs.512509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205280.1 NP_001192209.1 1, 2 138
XM_017026318.1 XP_016881807.1 1, 2 1191
GenBank mRNA BX112065.1 - 1, 2 138

View Full Product Details