Gene Transcripts

Gene Symbol
LELP1
Entrez Gene ID
149018
Gene Name
late cornified envelope like proline rich 1
Gene Aliases
Chromosome Location
Chr. 1: 153203430 - 153205125 on Build GRCh38
UniGene ID
Hs.125785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010857.2 NP_001010857.1 2 501
GenBank mRNA BC087854.1 - 2 441
DB069578.1 - 2 501
DB086762.1 - 2 501

View Full Product Details