Gene Transcripts

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
Chromosome Location
Chr. 20: 50586108 - 50692004 on Build GRCh38
UniGene ID
Hs.372578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290268.1 NP_001277197.1 21, 22 3145
NM_080829.3 NP_543019.2 21, 22 3078
NR_110890.1 21, 22 3304
XM_005260294.3 XP_005260351.1 22, 23 3142
XM_006723713.3 XP_006723776.1 21, 22 3190
XM_011528578.2 XP_011526880.1 22, 23 3170
XM_011528579.2 XP_011526881.1 22, 23 3308
XM_011528580.2 XP_011526882.1 22, 23 3362
XM_011528581.2 XP_011526883.1 22, 23 3346
XM_011528585.2 XP_011526887.1 11, 12 2493
XM_017027681.1 XP_016883170.1 13, 14 2138
GenBank mRNA AK299337.1 - 21, 22 2994
CA389799.1 - 2, 3 130
CD558804.1 - 2, 3 172

View Full Product Details