Gene Transcripts

Gene Symbol
SAMD14
Entrez Gene ID
201191
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
Chromosome Location
Chr. 17: 50111309 - 50129882 on Build GRCh38
UniGene ID
Hs.567769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257359.1 NP_001244288.1 4, 5 870
NM_174920.3 NP_777580.1 4, 5 870
XM_017024322.1 XP_016879811.1 3, 4 1254
XM_017024323.1 XP_016879812.1 5, 6 1037
XM_017024324.1 XP_016879813.1 4, 5 1725
GenBank mRNA AK094209.1 - 4, 5 801
AK095809.1 - 1, 2 171
AK225767.1 - 4, 5 801
AK291482.1 - 4, 5 797
AY148482.1 - 4, 5 791
BC047081.1 - 3, 4 524
BC136485.1 - 4, 5 783 9020474
BC141822.1 - 4, 5 810 40148162
BC142728.1 - 4, 5 767 40148957

View Full Product Details