Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145008.2 | NP_659445.1 | 4 | 686 | |
XM_017017318.1 | XP_016872807.1 | 4 | 2892 | ||
GenBank mRNA | AB098738.1 | - | 4 | 686 | |
AK054775.1 | - | 4 | 686 | ||
AK054967.1 | - | 3 | 1475 | ||
AK124577.1 | - | 4 | 2965 | ||
BC029228.1 | - | 3 | 862 | ||
BC064995.1 | - | 3 | 602 | ||
BC104781.1 | - | 4 | 541 | 8143784 | |
BC111012.1 | - | 3 | 876 | ||
BC112119.1 | - | 4 | 541 | 8327587 |