Gene Transcripts

Gene Symbol
CASC2
Entrez Gene ID
255082
Gene Name
cancer susceptibility candidate 2 (non-protein coding)
Gene Aliases
C10orf5
Chromosome Location
Chr. 10: 118046821 - 118210153 on Build GRCh38
UniGene ID
Hs.89387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026939.1 2 332
NR_026940.1 2 332
NR_026941.1 2 332
GenBank mRNA AJ344228.1 - 2 145
AJ535621.1 - 2 145
AJ535622.1 - 2 159
AK055184.1 - 2 159
AY226450.1 - 2 148
AY226451.1 - 2 148
BC036393.1 - 2 334
BC112306.1 - 1 56
BC112308.1 - 1 56
BC112326.1 - 2 152

View Full Product Details