Gene Transcripts
- Gene Symbol
- FRG1BP
- Entrez Gene ID
- 284802
- Gene Name
- FSHD region gene 1 family member B, pseudogene
- Gene Aliases
- C20orf80, FRG1B, bA348I14.2
- Chromosome Location
- Chr. 20: 30377203 - 30399331 on Build GRCh38
- UniGene ID
- Hs.653099
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC062743.1 |
- |
2 |
368 |
|
- Gene Symbol
- FRG1HP
- Entrez Gene ID
- 100132352
- Gene Name
- FSHD region gene 1 family member H, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 9: 40992337 - 41014139 on Build GRCh38
- UniGene ID
- Hs.231861
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034006.2 |
|
1 |
365 |
|
GenBank mRNA |
CN268684.1 |
- |
1 |
365 |
|
- Gene Symbol
- LOC102725126
- Entrez Gene ID
- 102725126
- Gene Name
- FSHD region gene 1 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.231861
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136306.1 |
|
1 |
440 |
|
GenBank mRNA |
AK311167.1 |
- |
1 |
385 |
|
DA426321.1 |
- |
1 |
442 |
|
- Gene Symbol
- LOC283788
- Entrez Gene ID
- 283788
- Gene Name
- FSHD region gene 1 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. Un|NT_167213: 56349 - 99642 on Build GRCh38
- UniGene ID
- Hs.657131
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027436.2 |
|
1 |
443 |
|
GenBank mRNA |
AK055615.1 |
- |
1 |
443 |
|
View Full Product Details