Gene Transcripts
- Gene Symbol
- FAM86B1
- Entrez Gene ID
- 85002
- Gene Name
- family with sequence similarity 86 member B1
- Gene Aliases
- Chromosome Location
- Chr. 8: 12182104 - 12194118 on Build GRCh38
- UniGene ID
- Hs.675799
- Species
- Homo sapiens
- Gene Symbol
- FAM86B2
- Entrez Gene ID
- 653333
- Gene Name
- family with sequence similarity 86 member B2
- Gene Aliases
- Chromosome Location
- Chr. 8: 12424419 - 12436418 on Build GRCh38
- UniGene ID
- Hs.458413
- Species
- Homo sapiens
- Gene Symbol
- FAM86C1
- Entrez Gene ID
- 55199
- Gene Name
- family with sequence similarity 86 member C1
- Gene Aliases
- FAM86C
- Chromosome Location
- Chr. 11: 71787511 - 71801234 on Build GRCh38
- UniGene ID
- Hs.567527
- Species
- Homo sapiens
- Gene Symbol
- FAM86DP
- Entrez Gene ID
- 692099
- Gene Name
- family with sequence similarity 86, member D, pseudogene
- Gene Aliases
- FAM86D
- Chromosome Location
- Chr. 3: 75421552 - 75435115 on Build GRCh38
- UniGene ID
- Hs.591652
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024241.1 |
|
8 |
1741 |
|
GenBank mRNA |
BC043342.2 |
- |
7 |
1465 |
|
- Gene Symbol
- FAM86FP
- Entrez Gene ID
- 653113
- Gene Name
- family with sequence similarity 86, member A pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 12: 8231049 - 8242946 on Build GRCh38
- UniGene ID
- Hs.661749
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024254.1 |
|
5 |
1246 |
|
View Full Product Details