Gene Transcripts

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
Chromosome Location
Chr. 10: 26938195 - 27100498 on Build GRCh38
UniGene ID
Hs.361041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015928.1 XP_016871417.1 39 6969
XM_017015929.1 XP_016871418.1 40 6957

View Full Product Details