Gene Transcripts

Gene Symbol
SMCO2
Entrez Gene ID
341346
Gene Name
single-pass membrane protein with coiled-coil domains 2
Gene Aliases
C12orf70
Chromosome Location
Chr. 12: 27446482 - 27506911 on Build GRCh38
UniGene ID
Hs.282121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145010.1 NP_001138482.1 6, 7 780
XM_011520634.2 XP_011518936.1 6, 7 958
XM_011520635.2 XP_011518937.1 4, 5 457
XM_017019254.1 XP_016874743.1 4, 5 714
XM_017019255.1 XP_016874744.1 3, 4 564

View Full Product Details