Gene Transcripts
- Gene Symbol
- FAM99A
- Entrez Gene ID
- 387742
- Gene Name
- family with sequence similarity 99 member A (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. 11: 1665599 - 1667856 on Build GRCh38
- UniGene ID
- Hs.567908
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026643.1 |
|
3 |
623 |
|
GenBank mRNA |
AK124823.1 |
- |
1 |
1227 |
|
CR627417.1 |
- |
3 |
650 |
|
- Gene Symbol
- FAM99B
- Entrez Gene ID
- 100132464
- Gene Name
- family with sequence similarity 99 member B (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. 11: 1683270 - 1685629 on Build GRCh38
- UniGene ID
- Hs.117797
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026642.1 |
|
3 |
649 |
|
View Full Product Details