Gene Transcripts

Gene Symbol
TBC1D28
Entrez Gene ID
254272
Gene Name
TBC1 domain family member 28
Gene Aliases
Chromosome Location
Chr. 17: 18634434 - 18644431 on Build GRCh38
UniGene ID
Hs.434123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017024418.1 XP_016879907.1 1 151
XM_017024420.1 XP_016879909.1 1 1474

View Full Product Details