Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004316.2 | NP_001004316.2 | 13 | 1885 | |
XM_005247441.3 | XP_005247498.1 | 13 | 3697 | ||
XM_006713631.3 | XP_006713694.1 | 13 | 2409 | ||
XM_006713636.2 | XP_006713699.1 | 8 | 1105 | ||
XM_011512803.2 | XP_011511105.1 | 12 | 2109 | ||
XM_011512804.2 | XP_011511106.1 | 13 | 3930 | ||
XM_011512809.1 | XP_011511111.1 | 10 | 1516 | ||
XM_011512813.2 | XP_011511115.1 | 8 | 1189 | ||
XM_011512814.2 | XP_011511116.1 | 8 | 1097 | ||
XM_017006374.1 | XP_016861863.1 | 9 | 1293 | ||
XM_017006375.1 | XP_016861864.1 | 9 | 1284 | ||
GenBank mRNA | AK131473.1 | - | 14 | 2194 |