Gene Transcripts
- Gene Symbol
- LINC00863
- Entrez Gene ID
- 439994
- Gene Name
- long intergenic non-protein coding RNA 863
- Gene Aliases
- Chromosome Location
- Chr. 10: 87342411 - 87357302 on Build GRCh38
- UniGene ID
- Hs.651188
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135292.1 |
|
5 |
1258 |
|
GenBank mRNA |
BC082979.1 |
- |
5 |
1063 |
|
- Gene Symbol
- NUTM2A
- Entrez Gene ID
- 728118
- Gene Name
- NUT family member 2A
- Gene Aliases
- FAM22A
- Chromosome Location
- Chr. 10: 87221899 - 87235372 on Build GRCh38
- UniGene ID
- Hs.710565
- Species
- Homo sapiens
View Full Product Details