Gene Transcripts

Gene Symbol
GDPGP1
Entrez Gene ID
390637
Gene Name
GDP-D-glucose phosphorylase 1
Gene Aliases
C15orf58, VTC2
Chromosome Location
Chr. 15: 90234255 - 90242080 on Build GRCh38
UniGene ID
Hs.304673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013657.2 NP_001013679.2 4 830
NM_001322811.1 NP_001309740.1 5 949
GenBank mRNA AK130527.1 - 4 1416

View Full Product Details