Gene Transcripts

Gene Symbol
LRRC8C
Entrez Gene ID
84230
Gene Name
leucine rich repeat containing 8 family member C
Gene Aliases
AD158, FAD158
Chromosome Location
Chr. 1: 89615810 - 89719535 on Build GRCh38
UniGene ID
Hs.412836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032270.4 NP_115646.2 1 75
GenBank mRNA AK127881.1 - 1 12

View Full Product Details