Gene Transcripts

Gene Symbol
RGS7BP
Entrez Gene ID
401190
Gene Name
regulator of G-protein signaling 7 binding protein
Gene Aliases
R7BP
Chromosome Location
Chr. 5: 64504565 - 64614661 on Build GRCh38
UniGene ID
Hs.657133
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029875.2 NP_001025046.1 5 1338
NM_001271890.1 NP_001258819.1 3 452
NM_001271891.1 NP_001258820.1 3 450
NR_073511.1 3 452
XM_005248502.3 XP_005248559.1 5 2718
GenBank mRNA AK294808.1 - 4 443
AK296443.1 - 5 986
AK296494.1 - 3 420
BC140707.1 - 5 684 9021575
BX640900.1 - 5 576
DA211241.1 - 3 453
DA417118.1 - 3 420

View Full Product Details