Gene Transcripts

Gene Symbol
SNX30
Entrez Gene ID
401548
Gene Name
sorting nexin family member 30
Gene Aliases
ATG24A
Chromosome Location
Chr. 9: 112750817 - 112875847 on Build GRCh38
UniGene ID
Hs.522350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012994.1 NP_001013012.1 3 514
XM_011518691.2 XP_011516993.1 3 551
XM_017014716.1 XP_016870205.1 3 582
GenBank mRNA BG699098.1 - 3 535

View Full Product Details