Gene Transcripts

Gene Symbol
OPA3
Entrez Gene ID
80207
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
Gene Aliases
MGA3
Chromosome Location
Chr. 19: 45527767 - 45584864 on Build GRCh38
UniGene ID
Hs.466945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025136.3 NP_079412.1 2 3389
XM_006723403.3 XP_006723466.1 3 3351

View Full Product Details