Gene Transcripts

Gene Symbol
RUFY3
Entrez Gene ID
22902
Gene Name
RUN and FYVE domain containing 3
Gene Aliases
RIPX, SINGAR1, ZFYVE30
Chromosome Location
Chr. 4: 70704204 - 70808622 on Build GRCh38
UniGene ID
Hs.740904
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037442.3 NP_001032519.1 16, 17 2376
NM_001291993.1 NP_001278922.1 17, 18 1768
XM_005265656.2 XP_005265713.1 17, 18 2414
XM_005265658.1 XP_005265715.1 16, 17 1690
XM_011531750.2 XP_011530052.1 17, 18 2012
GenBank mRNA AK302637.1 - 17, 18 1644

View Full Product Details