Gene Transcripts
- Gene Symbol
- SPDYE2
- Entrez Gene ID
- 441273
- Gene Name
- speedy/RINGO cell cycle regulator family member E2
- Gene Aliases
- SPDYB2-L1
- Chromosome Location
- Chr. 7: 102551232 - 102565227 on Build GRCh38
- UniGene ID
- Hs.656428
- Species
- Homo sapiens
- Gene Symbol
- POLR2J4
- Entrez Gene ID
- 84820
- Gene Name
- RNA polymerase II subunit J4, pseudogene
- Gene Aliases
- RPB11-phi
- Chromosome Location
- Chr. 7: 43940895 - 44019194 on Build GRCh38
- UniGene ID
- Hs.657028
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC006435.1 |
- |
1 |
359 |
|
BC017341.2 |
- |
4 |
477 |
|
- Gene Symbol
- SPDYE1
- Entrez Gene ID
- 285955
- Gene Name
- speedy/RINGO cell cycle regulator family member E1
- Gene Aliases
- Ringo1, SPDYE, WBSCR19
- Chromosome Location
- Chr. 7: 43997855 - 44011040 on Build GRCh38
- UniGene ID
- Hs.645483
- Species
- Homo sapiens
- Gene Symbol
- SPDYE18
- Entrez Gene ID
- 100505767
- Gene Name
- speedy/RINGO cell cycle regulator family member E18
- Gene Aliases
- Chromosome Location
- Chr. 7: 77051689 - 77060537 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- SPDYE2B
- Entrez Gene ID
- 100310812
- Gene Name
- speedy/RINGO cell cycle regulator family member E2B
- Gene Aliases
- SPDYE2L
- Chromosome Location
- Chr. 7: 102650325 - 102663525 on Build GRCh38
- UniGene ID
- Hs.433879
- Species
- Homo sapiens
- Gene Symbol
- SPDYE6
- Entrez Gene ID
- 729597
- Gene Name
- speedy/RINGO cell cycle regulator family member E6
- Gene Aliases
- Chromosome Location
- Chr. 7: 102345752 - 102356444 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details