Gene Transcripts
- Gene Symbol
- HCCAT5
- Entrez Gene ID
- 283902
- Gene Name
- hepatocellular carcinoma associated transcript 5 (non-protein coding)
- Gene Aliases
- FJ222407, HTA
- Chromosome Location
- Chr. 16: 73092349 - 73093774 on Build GRCh38
- UniGene ID
- Hs.382776
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027756.1 |
|
1, 2 |
636 |
|
GenBank mRNA |
FJ222407.1 |
- |
1, 2 |
636 |
|
View Full Product Details