Gene Transcripts

Gene Symbol
GOLGA6L6
Entrez Gene ID
727832
Gene Name
golgin A6 family-like 6
Gene Aliases
Chromosome Location
Chr. 15: 20531856 - 20541800 on Build GRCh38
UniGene ID
Hs.569472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145004.2 NP_001138476.2 2, 3 389

View Full Product Details