Gene Transcripts

Gene Symbol
GIPC3
Entrez Gene ID
126326
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
C19orf64, DFNB15, DFNB72, DFNB95
Chromosome Location
Chr. 19: 3585531 - 3593541 on Build GRCh38
UniGene ID
Hs.266873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133261.2 NP_573568.1 6 2866
XM_005259492.3 XP_005259549.1 6 2919
GenBank mRNA AK098244.1 - 1 582
BX648927.1 - 1 1812

View Full Product Details