Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177984.2 | NP_001171455.1 | 26 | 8803 | |
NM_014191.3 | NP_055006.1 | 27 | 8926 | ||
XM_006719556.3 | XP_006719619.1 | 27 | 9517 | ||
XM_011538650.2 | XP_011536952.1 | 27 | 8945 | ||
XM_011538651.2 | XP_011536953.1 | 27 | 8910 | ||
XM_017019794.1 | XP_016875283.1 | 27 | 8923 | ||
XM_017019795.1 | XP_016875284.1 | 26 | 8819 | ||
GenBank mRNA | AK091315.1 | - | 1 | 230 |