Gene Transcripts

Gene Symbol
ASPSCR1
Entrez Gene ID
79058
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
Chromosome Location
Chr. 17: 81977550 - 82018929 on Build GRCh38
UniGene ID
Hs.298351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090682.1 - 1 1188
AK126967.1 - 1 3483
AK127159.1 - 1 395

View Full Product Details