Gene Transcripts
- Gene Symbol
- ASPSCR1
- Entrez Gene ID
- 79058
- Gene Name
- ASPSCR1, UBX domain containing tether for SLC2A4
- Gene Aliases
- ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
- Chromosome Location
- Chr. 17: 81977550 - 82018929 on Build GRCh38
- UniGene ID
- Hs.298351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090682.1 |
- |
1 |
1243 |
|
AK126967.1 |
- |
1 |
3538 |
|
AK127159.1 |
- |
1 |
450 |
|
View Full Product Details