Gene Transcripts

Gene Symbol
LOC100130468
Entrez Gene ID
100130468
Gene Name
uncharacterized LOC100130468
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.662509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK126528.1 - 1 2571

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
Chromosome Location
Chr. 18: 10454628 - 10488701 on Build GRCh38
UniGene ID
Hs.293274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011525603.1 XP_011523905.1 4 4348

View Full Product Details