Gene Transcripts

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location
Chr. 3: 35638981 - 35798933 on Build GRCh38
UniGene ID
Hs.475902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267616.1 NP_001254545.1 1 489
GenBank mRNA BC041385.1 - 1 77

View Full Product Details