Gene Transcripts
- Gene Symbol
- WDR90
- Entrez Gene ID
- 197335
- Gene Name
- WD repeat domain 90
- Gene Aliases
- C16orf15, C16orf16, C16orf17, C16orf18, C16orf19
- Chromosome Location
- Chr. 16: 649363 - 667833 on Build GRCh38
- UniGene ID
- Hs.511903
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB067511.1 |
- |
12 |
2896 |
|
View Full Product Details