Gene Transcripts
- Gene Symbol
- SIRPG
- Entrez Gene ID
- 55423
- Gene Name
- signal regulatory protein gamma
- Gene Aliases
- CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1
- Chromosome Location
- Chr. 20: 1629152 - 1686516 on Build GRCh38
- UniGene ID
- Hs.590883
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124224.1 |
- |
1 |
765 |
|
View Full Product Details