Gene Transcripts

Gene Symbol
TEN1
Entrez Gene ID
100134934
Gene Name
TEN1 CST complex subunit
Gene Aliases
C17orf106
Chromosome Location
Chr. 17: 75979217 - 76000586 on Build GRCh38
UniGene ID
Hs.593566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113324.2 NP_001106795.2 4 633
GenBank mRNA BC047782.1 - 4 630
BX648576.1 - 4 599
FJ790671.1 - 3 329

Gene Symbol
TEN1-CDK3
Entrez Gene ID
100529145
Gene Name
TEN1-CDK3 readthrough (NMD candidate)
Gene Aliases
C17orf106-CDK3
Chromosome Location
Chr. 17: 75979217 - 76005999 on Build GRCh38
UniGene ID
Hs.706766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037709.1 4 633
GenBank mRNA AK097104.1 - 4 590
BX647274.1 - 4 619

View Full Product Details