Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144663.1 | NP_001138135.1 | 16 | 2387 | |
NM_004063.3 | NP_004054.3 | 16 | 2386 | ||
XM_011516790.1 | XP_011515092.1 | 15 | 2145 | ||
GenBank mRNA | AK300638.1 | - | 8 | 1265 | |
AK301001.1 | - | 12 | 1712 | ||
AK312873.1 | - | 16 | 2387 | ||
BC112013.1 | - | 16 | 2359 | 8327481 | |
BC113464.1 | - | 16 | 2347 | 8322516 | |
U07969.1 | - | 16 | 2348 | ||
X83228.1 | - | 16 | 2353 |