Gene Transcripts

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
Chromosome Location
Chr. 1: 149928651 - 149936898 on Build GRCh38
UniGene ID
Hs.425144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145862.1 NP_001139334.1 14 1590
NM_181873.3 NP_870988.2 13 1332
XM_006711137.1 XP_006711200.1 13 1309
XM_011509099.2 XP_011507401.1 14 1436
GenBank mRNA AK097000.1 - 13 1309
AK296094.1 - 12 1262
AK298775.1 - 14 1559
AK304691.1 - 13 1359
BC032655.1 - 7 664
BC064372.1 - 12 1551
BC094756.1 - 10 2617
BC100818.2 - 13 1250
BC100819.2 - 13 1250
BC100820.1 - 13 1250
BC100821.2 - 13 1203
U78556.1 - 12 1513
U78557.1 - 8 918

View Full Product Details