Gene Transcripts
- Gene Symbol
- MORF4L1
- Entrez Gene ID
- 10933
- Gene Name
- mortality factor 4 like 1
- Gene Aliases
- Eaf3, FWP006, HsT17725, MEAF3, MORFRG15, MRG15, S863-6
- Chromosome Location
- Chr. 15: 78872781 - 78897739 on Build GRCh38
- UniGene ID
- Hs.374503
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001265604.1 |
NP_001252533.1
|
2, 3 |
266 |
|
NM_001265605.1 |
NP_001252534.1
|
2, 3 |
166 |
|
NM_006791.3 |
NP_006782.1
|
2, 3 |
313 |
|
NM_206839.2 |
NP_996670.1
|
2, 3 |
313 |
|
XM_017021872.1 |
XP_016877361.1
|
2, 3 |
278 |
|
GenBank mRNA |
AF070664.1 |
- |
2, 3 |
120 |
|
AF100615.1 |
- |
2, 3 |
207 |
|
AF109188.1 |
- |
2, 3 |
102 |
|
AF161546.1 |
- |
2, 3 |
99 |
|
AF167173.1 |
- |
2, 3 |
115 |
|
AF218011.1 |
- |
2, 3 |
218 |
|
AK095823.1 |
- |
3, 4 |
715 |
|
AK098485.1 |
- |
2, 3 |
233 |
|
AK296650.1 |
- |
2, 3 |
204 |
|
AK300789.1 |
- |
2, 3 |
250 |
|
AK310181.1 |
- |
1, 2 |
51 |
|
AK310592.1 |
- |
2, 3 |
112 |
|
AK310702.1 |
- |
2, 3 |
121 |
|
AL137697.1 |
- |
2, 3 |
107 |
|
AY148481.1 |
- |
2, 3 |
207 |
|
BC002936.2 |
- |
2, 3 |
194 |
3955120 |
BC022845.2 |
- |
2, 3 |
194 |
2988803 |
BC067826.1 |
- |
2, 3 |
89 |
4824022 |
BC141840.1 |
- |
2, 3 |
121 |
8860303 |
BG771854.1 |
- |
2, 3 |
323 |
|
BX382045.2 |
- |
2, 3 |
110 |
|
DB455429.1 |
- |
2, 3 |
166 |
|
View Full Product Details