Gene Transcripts

Gene Symbol
UBE2C
Entrez Gene ID
11065
Gene Name
ubiquitin conjugating enzyme E2 C
Gene Aliases
UBCH10, dJ447F3.2
Chromosome Location
Chr. 20: 45812576 - 45816957 on Build GRCh38
UniGene ID
Hs.93002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007019.3 NP_008950.1 3, 4 345
NM_181801.3 NP_861517.1 3, 4 382
GenBank mRNA AK311949.1 - 3, 4 312
BC007656.2 - 3, 4 268 3345575
BC016292.1 - 3, 4 274 3910286
BC028161.1 - 2, 3 884
BC050736.1 - 3, 4 295 6503915
BF974975.1 - 401
BI858659.1 - 3, 4 382
BM747513.1 - 3, 4 300
BP227692.1 - 3, 4 372
BT007300.1 - 3, 4 225
BT020165.1 - 3, 4 225
BT020166.1 - 3, 4 225
CR456988.1 - 3, 4 225
DB062573.1 - 3, 4 346
U73379.1 - 3, 4 265

View Full Product Details