Gene Transcripts

Gene Symbol
PPP1R27
Entrez Gene ID
116729
Gene Name
protein phosphatase 1 regulatory subunit 27
Gene Aliases
DYSFIP1
Chromosome Location
Chr. 17: 81833492 - 81835050 on Build GRCh38
UniGene ID
Hs.454720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007533.3 NP_001007534.1 1 186
GenBank mRNA AF434846.1 - 1 164
AK096888.1 - 1 186
BC093707.1 - 1 129 7939552
BC112094.1 - 1 129 8327562
BX281734.1 - 1 169
DA894509.1 - 1 186

View Full Product Details