Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010846.2 | NP_001010846.1 | 3 | 822 | |
NR_135169.1 | 3 | 822 | |||
XM_005244891.4 | XP_005244948.1 | 3 | 825 | ||
XM_011509163.2 | XP_011507465.1 | 3 | 831 | ||
GenBank mRNA | AK074067.1 | - | 3 | 241 | |
AK126006.1 | - | 1 | 92 | ||
BC117210.1 | - | 3 | 822 | 40125761 | |
BC143765.1 | - | 3 | 821 | 9052279 | |
CR936667.1 | - | 3 | 270 |