Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040022.1 | NP_001035111.1 | 5 | 1253 | |
NM_001040023.1 | NP_001035112.1 | 4 | 1161 | ||
NM_080792.2 | NP_542970.1 | 5 | 920 | ||
XM_005260669.3 | XP_005260726.1 | 5 | 1303 | ||
XM_005260670.3 | XP_005260727.1 | 5 | 919 | ||
XM_011529173.2 | XP_011527475.1 | 4 | 908 | ||
GenBank mRNA | AB023430.1 | - | 5 | 920 | |
AK290776.1 | - | 5 | 1026 | ||
AK298243.1 | - | 5 | 871 | ||
AK312521.1 | - | 4 | 974 | ||
BC026692.1 | - | 5 | 925 | 3916836 | |
BC033092.1 | - | 4 | 913 | 4589298 | |
BC038510.2 | - | 4 | 1158 | 5261789 | |
BC075849.1 | - | 5 | 1099 | 4589732 | |
D86043.1 | - | 4 | 892 | ||
Y10375.1 | - | 4 | 902 |