Gene Transcripts

Gene Symbol
LEKR1
Entrez Gene ID
389170
Gene Name
leucine, glutamate and lysine rich 1
Gene Aliases
Chromosome Location
Chr. 3: 156825228 - 157047659 on Build GRCh38
UniGene ID
Hs.478048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193283.1 NP_001180212.1 1 491
XM_005247441.3 XP_005247498.1 1 1622
XM_006713631.3 XP_006713694.1 1 563
XM_011512803.2 XP_011511105.1 1 563
XM_011512804.2 XP_011511106.1 1 1622
XM_011512806.2 XP_011511108.1 1 563
XM_011512807.2 XP_011511109.1 1 563
XM_011512808.2 XP_011511110.1 1 563
XM_011512811.2 XP_011511113.1 1 563
XM_011512815.2 XP_011511117.1 1 563
XM_011512816.2 XP_011511118.1 1 563
GenBank mRNA AK094480.1 - 1 564
BC041967.1 - 1 491

View Full Product Details