Gene Transcripts

Gene Symbol
CX3CR1
Entrez Gene ID
1524
Gene Name
C-X3-C motif chemokine receptor 1
Gene Aliases
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
Chromosome Location
Chr. 3: 39263494 - 39281735 on Build GRCh38
UniGene ID
Hs.78913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171171.1 NP_001164642.1 2 1085
NM_001171172.1 NP_001164643.1 2 1045
NM_001171174.1 NP_001164645.1 2 981
NM_001337.3 NP_001328.1 2 937
GenBank mRNA AK225764.1 - 2 938
AK307911.1 - 3 737
BC028078.1 - 2 921 5216452
U20350.1 - 2 932
U28934.1 - 2 890

View Full Product Details