Gene Transcripts

Gene Symbol
ANKFN1
Entrez Gene ID
162282
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 56110935 - 56517016 on Build GRCh38
UniGene ID
Hs.673040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153228.2 NP_694960.2 12 1413
XM_006721728.3 XP_006721791.1 11 1636
XM_011524428.2 XP_011522730.1 12 1413
XM_011524429.2 XP_011522731.1 12 1570
XM_011524430.2 XP_011522732.1 13 1536
XM_011524431.2 XP_011522733.1 12 1468
XM_011524433.2 XP_011522735.1 6 826
XM_011524434.2 XP_011522736.1 3 273
XM_017024263.1 XP_016879752.1 14 1630
XM_017024264.1 XP_016879753.1 14 2615
XM_017024265.1 XP_016879754.1 14 1824
XM_017024266.1 XP_016879755.1 12 1585
XM_017024267.1 XP_016879756.1 11 1437
XM_017024268.1 XP_016879757.1 13 2521
XM_017024269.1 XP_016879758.1 11 1710
XM_017024270.1 XP_016879759.1 14 1628
GenBank mRNA AK095654.1 - 12 1413

View Full Product Details