Gene Transcripts

Gene Symbol
WDR90
Entrez Gene ID
197335
Gene Name
WD repeat domain 90
Gene Aliases
C16orf15, C16orf16, C16orf17, C16orf18, C16orf19
Chromosome Location
Chr. 16: 649363 - 667833 on Build GRCh38
UniGene ID
Hs.511903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145294.4 NP_660337.3 4, 5 441
XM_017023023.1 XP_016878512.1 4, 5 441
GenBank mRNA AB067511.1 - 3, 4 829
AK091062.1 - 4, 5 441
BC121185.1 - 4, 5 451
BC121186.1 - 4, 5 451

View Full Product Details