Gene Transcripts
- Gene Symbol
- DNAJC13
- Entrez Gene ID
- 23317
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C13
- Gene Aliases
- PARK21, RME8
- Chromosome Location
- Chr. 3: 132417527 - 132539032 on Build GRCh38
- UniGene ID
- Hs.12707
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015268.3 |
NP_056083.3
|
55, 56 |
6700 |
|
XM_005247245.4 |
XP_005247302.1
|
56, 57 |
6897 |
|
XM_017006036.1 |
XP_016861525.1
|
42, 43 |
5157 |
|
GenBank mRNA |
AB014578.2 |
- |
55, 56 |
6749 |
|
AK023168.1 |
- |
24, 25 |
3128 |
|
AK125330.1 |
- |
32, 33 |
4084 |
|
AK127112.1 |
- |
39, 40 |
4788 |
|
AY779857.1 |
- |
55, 56 |
6700 |
|
BC000164.3 |
- |
8, 9 |
1063 |
|
BC009630.1 |
- |
10, 11 |
1183 |
|
BC040638.2 |
- |
5, 6 |
751 |
|
BC065756.1 |
- |
4, 5 |
459 |
|
BC105580.1 |
- |
3, 4 |
276 |
|
BC151246.1 |
- |
56, 57 |
6770 |
9007477 |
View Full Product Details