Gene Transcripts

Gene Symbol
QPRT
Entrez Gene ID
23475
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
HEL-S-90n, QPRTase
Chromosome Location
Chr. 16: 29670588 - 29698699 on Build GRCh38
UniGene ID
Hs.513484
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014298.4 NP_055113.2 2 694
XM_005255223.3 XP_005255280.2 3 777
XM_017023101.1 XP_016878590.1 3 777
GenBank mRNA BC005060.1 - 2 531
BC010033.2 - 2 523 3506460
BC018910.2 - 2 548 3959973
BT007231.1 - 1 504
EU794638.1 - 2 523
KJ534923.1 - 1 504

View Full Product Details