Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173854.5 | NP_776253.3 | 9 | 2222 | |
XM_005245069.2 | XP_005245126.1 | 9 | 1898 | ||
GenBank mRNA | AJ514402.1 | - | 8 | 1309 | |
AK093254.1 | - | 1 | 1243 | ||
AK125075.1 | - | 8 | 983 | ||
AK128069.1 | - | 8 | 1475 | ||
AK314732.1 | - | 8 | 1380 | ||
AL831974.1 | - | 9 | 2058 | ||
AL832001.1 | - | 9 | 2058 | ||
AL832362.1 | - | 9 | 2058 | ||
BC136303.1 | - | 8 | 1703 | 9020290 | |
BX648979.1 | - | 9 | 2096 |