Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153613.2 | NP_705841.2 | 4 | 591 | |
XM_006720454.2 | XP_006720517.1 | 4 | 593 | ||
XM_011521422.1 | XP_011519724.1 | 4 | 573 | ||
GenBank mRNA | AF529233.1 | - | 4 | 442 | |
AF542964.1 | - | 4 | 675 | ||
AK290273.1 | - | 4 | 562 | ||
AK308910.1 | - | 4 | 573 | ||
AL832237.1 | - | 4 | 588 | ||
AY734233.1 | - | 4 | 611 | ||
BC028209.1 | - | 2 | 147 | ||
BC048430.1 | - | 4 | 593 | ||
BC092463.1 | - | 4 | 588 | 30520144 | |
BC144236.1 | - | 4 | 519 |