Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167621.1 | NP_001161093.1 | 8 | 1184 | |
NM_013236.3 | NP_037368.1 | 9 | 1376 | ||
GenBank mRNA | AF086923.1 | - | 9 | 1328 | |
AF119662.1 | - | 9 | 1316 | ||
AK025734.1 | - | 9 | 1307 | ||
AK095309.1 | - | 9 | 1144 | ||
AK294348.1 | - | 8 | 1177 | ||
AL050282.1 | - | 9 | 1308 | ||
BC007508.2 | - | 9 | 1298 | 3030062 | |
CR456568.1 | - | 9 | 1161 | ||
CR457381.1 | - | 9 | 1110 |