Gene Transcripts

Gene Symbol
C1orf43
Entrez Gene ID
25912
Gene Name
chromosome 1 open reading frame 43
Gene Aliases
HSPC012, NICE-3, NS5ATP4, S863-3
Chromosome Location
Chr. 1: 154206701 - 154220797 on Build GRCh38
UniGene ID
Hs.287471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098616.2 NP_001092086.1 7 994
NM_001297717.1 NP_001284646.1 5 881
NM_001297718.1 NP_001284647.1 7 917
NM_001297720.1 NP_001284649.1 6 940
NM_015449.3 NP_056264.1 6 892
NM_138740.3 NP_620077.1 5 838
GenBank mRNA AF077036.1 - 6 712
AF109185.1 - 7 685
AF529365.1 - 7 604
AJ243663.1 - 5 660
AJ243664.1 - 6 660
AJ243665.1 - 6 497
AL117463.1 - 5 458
BC000152.2 - 7 803 2901006
BC001377.2 - 6 660 3051048
BC006514.1 - 6 729 2822327
BC014391.1 - 6 628 3544590
BC088353.1 - 5 701 6164281
CR457131.1 - 6 502
DQ908899.1 - 6 502
DR007097.1 - 5 721

View Full Product Details