Gene Transcripts

Gene Symbol
CFAP61
Entrez Gene ID
26074
Gene Name
cilia and flagella associated protein 61
Gene Aliases
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
Chromosome Location
Chr. 20: 20052514 - 20360714 on Build GRCh38
UniGene ID
Hs.729168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015585.3 NP_056400.3 17, 18 2054
XM_005260688.3 XP_005260745.1 17, 18 2032
XM_005260689.2 XP_005260746.1 17, 18 2017
XM_005260690.3 XP_005260747.1 16, 17 1957
XM_005260691.3 XP_005260748.1 16, 17 1954
XM_005260692.3 XP_005260749.1 16, 17 1939
XM_005260693.3 XP_005260750.1 17, 18 2032
XM_005260695.4 XP_005260752.1 12, 13 2343
XM_005260696.4 XP_005260753.1 8, 9 916
XM_011529211.1 XP_011527513.1 16, 17 1890
XM_011529212.2 XP_011527514.1 17, 18 2032
XM_017027786.1 XP_016883275.1 17, 18 1972
XM_017027787.1 XP_016883276.1 17, 18 1957
XM_017027788.1 XP_016883277.1 16, 17 1939
XM_017027789.1 XP_016883278.1 17, 18 2032
XM_017027790.1 XP_016883279.1 17, 18 2032
XM_017027791.1 XP_016883280.1 7, 8 814
XM_017027792.1 XP_016883281.1 8, 9 856
XM_017027793.1 XP_016883282.1 7, 8 823
XM_017027794.1 XP_016883283.1 7, 8 887
GenBank mRNA AK304174.1 - 17, 18 1957
BC028708.2 - 17, 18 2016 4825941

View Full Product Details